missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus Envelope Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56744
This item is not returnable.
View return policy
Description
West Nile Virus Envelope Polyclonal specifically detects West Nile Virus Envelope in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| West Nile Virus Envelope | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| West Nile Virus envelope glycoprotein E | |
| A synthetic peptide corresponding to 73-86 (CPTMGEAHNEKRAD) of West Nile virus envelope glycoprotein (NC_001563). | |
| 0.1 mg | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry 1 ug/ml, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24920798), Immunohistochemistry-Paraffin 1 ug/ml | |
| P06935 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction