missing translation for 'onlineSavingsMsg'
Learn More
Learn More
West Nile Virus Envelope Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£167.00 - £407.00
Specifications
| Applications | Western Blot, Immunohistochemistry |
|---|---|
| Form | Purified |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Isotype | IgG |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18780074
|
Novus Biologicals
NB100-56744SS |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18765923
|
Novus Biologicals
NB100-56744 |
0.1 mg |
£407.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
West Nile Virus Envelope Polyclonal specifically detects West Nile Virus Envelope in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| Western Blot, Immunohistochemistry | |
| Rabbit | |
| IgG |
| Purified | |
| RUO |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title