missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24662
This item is not returnable.
View return policy
Description
GLI-1 Polyclonal specifically detects GLI-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GLI-1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| GLI1 | |
| A portion of amino acids 940-990 of human GLI1 was used as the immunogen for this antibody. | |
| 0.1 mg | |
| Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| 2735 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human and rhesus monkey, 94% homologous in horse, 88% homologous in dog, and 76% homologous in cow, mouse and rat. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction