missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£177.00 - £405.00
Specifications
| Antigen | GLI-1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18110392
|
Novus Biologicals
NBP2-24662SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726173
|
Novus Biologicals
NBP2-24662 |
0.1 mg |
£405.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GLI-1 Polyclonal specifically detects GLI-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GLI-1 | |
| Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| GLI family zinc finger 1, Glioma-associated oncogene, glioma-associated oncogene family zinc finger 1, glioma-associated oncogene homolog 1 (zinc finger protein), GLIzinc finger protein GLI1, Oncogene GLI | |
| GLI1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human and rhesus monkey, 94% homologous in horse, 88% homologous in dog, and 76% homologous in cow, mouse and rat. |
| 1.0 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| 2735 | |
| A portion of amino acids 940-990 of human GLI1 was used as the immunogen for this antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title