missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Flu M1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56610
This item is not returnable.
View return policy
Description
Avian Flu M1 Polyclonal antibody specifically detects Avian Flu M1 in Virus samples. It is validated for Western Blot.
Specifications
| Avian Flu M1 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| The amino acid sequence used as immunogen is 100% homologous to duck H5N1 strain and 92% homologous to chicken H5N1 strain. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml | |
| Avian Influenza Matrix Protein 1 | |
| Amino acids 212-225 of avian flu matrix protein 1 were used as the immunogen. | |
| 0.1 mg | |
| Primary | |
| Virus, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction