missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Flu M1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £360.00
Specifications
| Antigen | Avian Flu M1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18093224
|
Novus Biologicals
NB100-56610SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18707223
|
Novus Biologicals
NB100-56610 |
0.1 mg |
£360.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Avian Flu Matrix Protein 1 Polyclonal specifically detects Avian Flu Matrix Protein 1 in Avian, Chicken samples. It is validated for Western Blot.Specifications
| Avian Flu M1 | |
| Western Blot 0.5-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| Amino acids 212-225 of avian flu matrix protein 1 were used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Virus, Chicken | |
| Avian Influenza Matrix Protein 1 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous to duck H5N1 strain and 92% homologous to chicken H5N1 strain. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title