missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 0.1% Peptone Water
Description
Diluent used in the preparation of food and water samples prior to further testing
- Diluent used in the preparation of food and water samples prior to further testing
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | United Kingdom |
| Product Type | Broth Tube |
| Sterility | Non-sterile |
| Target Organism Class | Bacteria |
| Test Type | Microbial Detection |
| Quantity | 10 x 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?