missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94081-0.02ml
This item is not returnable.
View return policy
Description
OAS3 Polyclonal antibody specifically detects OAS3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| OAS3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| (2-5')oligo(A) synthase 3,2'-5'oligoadenylate synthetase p100, 2-5A synthase 3, 2'-5'-oligoadenylate synthase 3, 2'-5'-oligoadenylate synthetase 3, 100kDa, P/OKcl.4, p100 OAS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-620 of human OAS3 (NP_006178.2). AASCYDHPCFLRGMGDPVQSWKGPGLPRAGCSGLGHPIQLDPNQKTPENSKSLNAVYPRAGSKPPSCPAPGPTGAASIVPSVPGMALDLSQIPTKELDRFIQDHLKPSPQFQEQVKKAIDIILRCLHENCVHKASRVSKGGSFGRGTDLRDGCDVELIIFLNCFTDYKDQGPRRAEILDEMRAQLESWWQDQVPSLSLQFPEQNVPEALQFQLVSTALKSWTDVSLLPAFDAVGQLSSGTKPNPQVYSRLLTSGCQEGEHKACFAELRRNFMNIRPVKLKNLILLVKHWYRQVAAQNKGKG | |
| 0.02 mL | |
| Immunology, Innate Immunity | |
| 4940 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction