missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £366.00
Specifications
| Antigen | OAS3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18630172
|
Novus Biologicals
NBP2-94081-0.02ml |
0.02 mL |
£150.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683392
|
Novus Biologicals
NBP2-94081-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OAS3 Polyclonal antibody specifically detects OAS3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| OAS3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity | |
| PBS (pH 7.3), 50% glycerol | |
| 4940 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| (2-5')oligo(A) synthase 3,2'-5'oligoadenylate synthetase p100, 2-5A synthase 3, 2'-5'-oligoadenylate synthase 3, 2'-5'-oligoadenylate synthetase 3, 100kDa, P/OKcl.4, p100 OAS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 320-620 of human OAS3 (NP_006178.2). AASCYDHPCFLRGMGDPVQSWKGPGLPRAGCSGLGHPIQLDPNQKTPENSKSLNAVYPRAGSKPPSCPAPGPTGAASIVPSVPGMALDLSQIPTKELDRFIQDHLKPSPQFQEQVKKAIDIILRCLHENCVHKASRVSKGGSFGRGTDLRDGCDVELIIFLNCFTDYKDQGPRRAEILDEMRAQLESWWQDQVPSLSLQFPEQNVPEALQFQLVSTALKSWTDVSLLPAFDAVGQLSSGTKPNPQVYSRLLTSGCQEGEHKACFAELRRNFMNIRPVKLKNLILLVKHWYRQVAAQNKGKG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title