Learn More
Insulin Antibody (IRDN/794), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45108-0.02MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Insulin Monoclonal specifically detects Insulin in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Monoclonal | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| 6 kDa | |
| Store at 4C. | |
| IgG1 κ |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
| INS | |
| Pig | |
| Purified |