Learn More
VPS34 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NB110-87320SS
 View more versions of this product
Product Code. 18707873
  • £189.00 / 25µL
Description
VPS34 Polyclonal specifically detects VPS34 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunofluorescence
Western Blot 1:1000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:100, Knockdown Validated
Affinity Purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
EC 2.7.1, EC 2.7.1.137, hVps34, MGC61518, phosphatidylinositol 3-kinase catalytic subunit type 3, Phosphatidylinositol 3-kinase p100 subunit, Phosphoinositide-3-kinase class 3, phosphoinositide-3-kinase, class 3, PI3K type 3, PI3-kinase type 3, PtdIns-3-kinase type 3, Vps34
Pig