missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS34 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£189.00 - £418.00
Specifications
| Dilution | Western Blot 1:1000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:100, Knockdown Validated |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Target Species | Pig |
| Gene Alias | EC 2.7.1, EC 2.7.1.137, hVps34, MGC61518, phosphatidylinositol 3-kinase catalytic subunit type 3, Phosphatidylinositol 3-kinase p100 subunit, Phosphoinositide-3-kinase class 3, phosphoinositide-3-kinase, class 3, PI3K type 3, PI3-kinase type 3, PtdIns-3-kinase type 3, Vps34 |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18707873
|
Novus Biologicals
NB110-87320SS |
£189.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18789563
|
Novus Biologicals
NB110-87320 |
£418.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
VPS34 Polyclonal specifically detects VPS34 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
| Western Blot 1:1000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:100, Knockdown Validated | |
| Polyclonal | |
| EC 2.7.1, EC 2.7.1.137, hVps34, MGC61518, phosphatidylinositol 3-kinase catalytic subunit type 3, Phosphatidylinositol 3-kinase p100 subunit, Phosphoinositide-3-kinase class 3, phosphoinositide-3-kinase, class 3, PI3K type 3, PI3-kinase type 3, PtdIns-3-kinase type 3, Vps34 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Pig | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title