Learn More
IRP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand:  Novus Biologicals NB100-1798SS
 View more versions of this product
Product Code. 18751454
  • £146.00 / 25µL
Description
IRP2 Polyclonal specifically detects IRP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Gel Supershift Assay.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IRP2
Polyclonal
Unconjugated
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
ACO3, EC 4.2.1.3, FLJ23381, IRE-BP 2, Iron regulatory protein 2, iron-responsive element binding protein 2, iron-responsive element-binding protein 2, IRP2IRP2AD
Rabbit
Affinity Purified
RUO
Primary
Mouse. Preliminary results have shown poor reactivity against human protein. Immunogen has 94% homology with human, primate and rat proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1.15 mg/mL
Western Blot 1:500-1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200-1:500, Gel Super Shift Assays reported in scientific literature (PMID 26752519), Gel Supershift Assay
Q811J3
IREB2
A synthetic peptide made to an internal portion of the murine IRP2 protein sequence (between residues 100-200). [UniProt Q811J3]
0.025 mL
HIF Target Genes, Hypoxia, Lipid and Metabolism
3658
Human, Mouse