missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£146.00 - £396.00
Specifications
| Antigen | IRP2 |
|---|---|
| Concentration | 1.15 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200-1:500, Gel Super Shift Assays reported in scientific literature (PMID 26752519), Gel Supershift Assay |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18751454
|
Novus Biologicals
NB100-1798SS |
0.025 mL |
£146.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18779133
|
Novus Biologicals
NB100-1798 |
0.1 mL |
£396.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRP2 Polyclonal specifically detects IRP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Gel Supershift Assay.Specifications
| IRP2 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200-1:500, Gel Super Shift Assays reported in scientific literature (PMID 26752519), Gel Supershift Assay | |
| Polyclonal | |
| Rabbit | |
| HIF Target Genes, Hypoxia, Lipid and Metabolism | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| ACO3, EC 4.2.1.3, FLJ23381, IRE-BP 2, Iron regulatory protein 2, iron-responsive element binding protein 2, iron-responsive element-binding protein 2, IRP2IRP2AD | |
| IREB2 | |
| IgG | |
| Affinity Purified | |
| Mouse. Preliminary results have shown poor reactivity against human protein. Immunogen has 94% homology with human, primate and rat proteins. |
| 1.15 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q811J3 | |
| 3658 | |
| A synthetic peptide made to an internal portion of the murine IRP2 protein sequence (between residues 100-200). [UniProt Q811J3] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title