missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk1 Antibody (2G1D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-1768SS
This item is not returnable.
View return policy
Description
Chk1 Monoclonal specifically detects Chk1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Chk1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:200 - 1:1000, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:250 | |
| Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| Mouse | |
| 56 kDa | |
| 0.025 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers, Tumor Suppressors | |
| 1111 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2G1D5 | |
| Unconjugated | |
| O14757 | |
| CHEK1 | |
| Purified recombinant fragment of Chk1 expressed in E. coli. [UniProt# O14757] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction