missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk1 Antibody (2G1D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£186.00 - £405.00
Specifications
| Antigen | Chk1 |
|---|---|
| Clone | 2G1D5 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:200 - 1:1000, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:250 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18705633
|
Novus Biologicals
NB100-1768SS |
0.025 mL |
£186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18000964
|
Novus Biologicals
NB100-1768 |
0.1 mL |
£405.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chk1 Monoclonal specifically detects Chk1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Chk1 | |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers, Tumor Suppressors | |
| O14757 | |
| 1111 | |
| Purified recombinant fragment of Chk1 expressed in E. coli. [UniProt# O14757] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2G1D5 | |
| Western Blot 1:200 - 1:1000, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:250 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Checkpoint, S. pombe, homolog of, 1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1 | |
| CHEK1 | |
| IgG1 | |
| Protein G purified | |
| 56 kDa |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts