accessibility menu, dialog, popup

UserName

CH3H2NSOHOO

CAS RN 98-33-9

IUPAC Name: 4-amino-3-methylbenzene-1-sulfonic acid
Synonyms: 4-amino-3-methylbenzenesulfonic acid
Molecular Weight (g/mol): 187.21
Molecular Formula: C7H9NO3S
InChi Key: WQTCZINVPXJNEL-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1N)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results