missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-methylbenzenesulfonic acid, 98%, Thermo Scientific™

Product Code. 11428707
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428707 250 g 250g
11418707 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428707 Supplier Thermo Scientific Alfa Aesar Supplier No. A19870.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-33-9
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00025198
InChI Key WQTCZINVPXJNEL-UHFFFAOYSA-N
Synonym 2-aminotoluene-5-sulfonic acid, benzenesulfonic acid, 4-amino-3-methyl, 4-amino-m-toluenesulfonic acid, 2-amino-5-toluenesulfonic acid, o-toluidine-m-sulfonic acid, 6-aminotoluene-3-sulfonic acid, 3-methylsulphanilic acid, unii-et833cb2ku, ccris 3412, 4-amino-m-toluenesulfonic acid so3h=1
PubChem CID 7384
IUPAC Name 4-amino-3-methylbenzenesulfonic acid
SMILES CC1=CC(=CC=C1N)S(O)(=O)=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 250 g
Formula Weight 187.22
Percent Purity 98%
Chemical Name or Material 4-Amino-3-methylbenzenesulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.