CAS RN 95382-33-5
CAS RN 95382-33-5
IUPAC Name:
magnesium(2+) bis(6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methanesulfinyl]-1H-1,3-benzodiazole)
Synonyms:
magnesium(2+) bis(omeprazole)
Molecular Weight (g/mol):
715.14
Molecular Formula:
C34H38MgN6O6S2
InChi Key:
ZAUIQBMEUIXDRJ-UHFFFAOYNA-N
SMILES:
[Mg++].COC1=CC=C2N=C(NC2=C1)S(=O)CC1=NC=C(C)C(OC)=C1C.COC1=CC=C2N=C(NC2=C1)S(=O)CC1=NC=C(C)C(OC)=C1C