missing translation for 'onlineSavingsMsg'
Learn More

Omeprazole Magnesium, TRC

Product Code. 17971855
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971855 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971855 Supplier Toronto Research Chemicals Supplier No. TRCO635065100MG
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 95382-33-5
Molecular Formula 2 C17 H18 N3 O3 S . Mg
Quantity 100 mg
Synonym Omeprazole magnesium, Magnesium bis[5-methoxy-2-[(RS)-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]sulfinyl]-1H-benzimidazol-1-ide]
InChI Formula InChI=1S/2C17H18N3O3S.Mg/c2*1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17;/h2*5-8H,9H2,1-4H3;/q2*-1;+2
SMILES [Mg+2].COc1ccc2[n-]c(nc2c1)S(=O)Cc3ncc(C)c(OC)c3C.COc4ccc5[n-]c(nc5c4)S(=O)Cc6ncc(C)c(OC)c6C
IUPAC Name magnesium;5-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]benzimidazol-1-ide
Molecular Weight (g/mol) 713.12
Formula Weight 712.1988
Chemical Name or Material Omeprazole magnesium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.