accessibility menu, dialog, popup

UserName

CAS RN 947249-19-6

BOOH3CCH3H3CH3CNNH2OCH3

CAS RN 947249-19-6

IUPAC Name: 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms: 3-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol): 250.11
Molecular Formula: C12H19BN2O3
InChi Key: OXNSEGHJPTXNPF-UHFFFAOYSA-N
SMILES: COC1=C(N)N=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results