accessibility menu, dialog, popup

UserName

CAS RN 942206-24-8

NNNClCl

CAS RN 942206-24-8

IUPAC Name: 6'-chloro-4-(6-chloropyridin-3-yl)-2,3'-bipyridine
Synonyms: 6'-chloro-4-(6-chloropyridin-3-yl)-2,3'-bipyridine
Molecular Weight (g/mol): 302.16
Molecular Formula: C15H9Cl2N3
InChi Key: CWMMDFDDGLQUBF-UHFFFAOYSA-N
SMILES: ClC1=NC=C(C=C1)C1=CC(=NC=C1)C1=CN=C(Cl)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results