missing translation for 'onlineSavingsMsg'
Learn More

6,6″-Dichloro-3,2':4',3″-terpyridine, 95%, Thermo Scientific™

Product Code. 11496500
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496500 500 mg 500mg
11486500 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496500 Supplier Thermo Scientific Alfa Aesar Supplier No. H27593.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942206-24-8
Molecular Formula C15H9Cl2N3
Molecular Weight (g/mol) 302.158
MDL Number MFCD09037443
InChI Key CWMMDFDDGLQUBF-UHFFFAOYSA-N
Synonym 6,6'-dichloro-3,2':4',3'-terpyridine, 2,4-bis 6-chloropyridin-3-yl pyridine, 6,6-dichloro-3,2':4',3-terpyridine, 6,6-dichloro-3,2':4',3'-terpyridine, 1∼6∼,3∼6∼-dichloro-1∼3∼,2∼2∼:2∼4∼,3∼3∼-terpyridine, 6'-chloro-4-6-chloropyridin-3-yl-2,3'-bipyridine
PubChem CID 42553048
IUPAC Name 2,4-bis(6-chloropyridin-3-yl)pyridine
SMILES C1=CC(=NC=C1C2=CC(=NC=C2)C3=CN=C(C=C3)Cl)Cl

Specifications

Quantity 500 mg
Formula Weight 302.16
Percent Purity 95%
Chemical Name or Material 6,6''-Dichloro-3,2':4',3''-terpyridine
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.