accessibility menu, dialog, popup

UserName

O

CAS RN 92-83-1

IUPAC Name: 9H-xanthene
Synonyms: xanthan gum dibenzo[a,e]pyran
Molecular Weight (g/mol): 182.22
Molecular Formula: C13H10O
InChi Key: GJCOSYZMQJWQCA-UHFFFAOYSA-N
SMILES: C1C2=CC=CC=C2OC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results