missing translation for 'onlineSavingsMsg'
Learn More

Xanthene 98%, Thermo Scientific™

Product Code. 10299543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10299543 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10299543 Supplier Thermo Scientific Acros Supplier No. 140950250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-83-1
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.222
MDL Number MFCD00005055
InChI Key GJCOSYZMQJWQCA-UHFFFAOYSA-N
Synonym xanthene, xanthan, 10h-9-oxaanthracene, dibenzo a,e pyran, dibenzopyran, tricyclic, 9h-xanthen, xanthane, 9h-xanthen-9-yl, acmc-209rhu
PubChem CID 7107
ChEBI CHEBI:10057
IUPAC Name 9H-xanthene
SMILES C1C2=CC=CC=C2OC3=CC=CC=C31

Specifications

Melting Point 100°C to 103°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 g
Beilstein 17, 73
Formula Weight 182.22
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Xanthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.