CAS RN 906673-24-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Crisaborole, TRC
CAS: 906673-24-3 Molecular Formula: C14H10BNO3 Molecular Weight (g/mol): 251.05 Synonym: ,4-((1-Hydroxy-1,3-dihydrobenzo[c][1,2]oxaborol-5-yl)oxy)benzonitrile,4-[(1,3-Dihydro-1-hydroxy-2,1-benzoxaborol-5-yl)oxy]benzonitrile,5-(4-Cyanophenoxy)-1,3-dihydro-1-hydroxy-[2,1]-benzoxaborole,5-(4-Cyanophenoxy)-2,3-dihydro-1-hydroxy-2,1-benzoxaborole,AN2728 IUPAC Name: 4-[(1-hydroxy-3H-2,1-benzoxaborol-5-yl)oxy]benzonitrile SMILES: OB1OCc2cc(Oc3ccc(cc3)C#N)ccc12
2
Crisaborole, MedChemExpress
MedChemExpress Crisaborole (AN-2728) is a potent inhibitor of PDE4 and cytokine release; inhibit PDE4 with an IC50 of 0.49 μM.