missing translation for 'onlineSavingsMsg'
Learn More

Crisaborole, MedChemExpress

Product Code. 16444328
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444328 10 mg 10mg
16425246 100 mg 100mg
16400657 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16444328 Supplier Medchem Express Supplier No. HY10978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Crisaborole (AN-2728) is a potent inhibitor of PDE4 and cytokine release; inhibit PDE4 with an IC50 of 0.49 μM.

Crisaborole (AN-2728) is a potent inhibitor of PDE4 and cytokine release

Specifications

Chemical Name or Material Crisaborole
CAS 906673-24-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H10BNO3
Quantity 10 mg
Synonym AN-2728 PF-06930164
Solubility Information DMSO : ≥ 100 mg/mL (398.33 mM)
SMILES N#CC1=CC=C(OC2=CC=C(B(O)OC3)C3=C2)C=C1
Molecular Weight (g/mol) 251.05
Formula Weight 251.05
Percent Purity 96.55%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.