accessibility menu, dialog, popup

UserName

CAS RN 90223-02-2

H3CNNNH2

CAS RN 90223-02-2

IUPAC Name: 2-methyl-2H-indazol-7-amine
Synonyms: 2-methylindazol-7-amine
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: KNZJAZZOKTUEHM-UHFFFAOYSA-N
SMILES: CN1C=C2C=CC=C(N)C2=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results