missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylumbelliferyl-alpha-L-iduronide Sodium Salt, Synthetic, Thermo Scientific Chemicals
Shop All Thermo Scientific ProductsSpecifications
Specifications
| CAS | 89157-94-8 |
| Physical Form | Powder |
| Quantity | 5 mg |
| Packaging | Plastic Bottle |
| Synonym | 4-methyl-2-oxo-2 h-1-benzopyran-7-yl alpha-l-idopyranosiduronic acid monosodium salt |
| Molecular Weight (g/mol) | 375.285 |
| Chemical Name or Material | 4-Methylumbelliferyl-alpha-L-iduronide Sodium Salt |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?