accessibility menu, dialog, popup

UserName

CAS RN 887268-08-8

FFFFFFF

CAS RN 887268-08-8

IUPAC Name: 2-fluoro-1,4-bis(trifluoromethyl)benzene
Synonyms: 2-fluoro-1,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol): 232.1
Molecular Formula: C8H3F7
InChi Key: KAGRJEBZBWYSJK-UHFFFAOYSA-N
SMILES: FC1=C(C=CC(=C1)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results