CAS RN 886498-13-1
CAS RN 886498-13-1
IUPAC Name:
1-[2-fluoro-5-(trifluoromethoxy)phenyl]methanamine
Synonyms:
1-[2-fluoro-5-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol):
209.14
Molecular Formula:
C8H7F4NO
InChi Key:
MBNLPGBRZXRZOM-UHFFFAOYSA-N
SMILES:
NCC1=C(F)C=CC(OC(F)(F)F)=C1
1
–
1
of
1
results
1
2-Fluoro-5-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™
CAS: 886498-13-1 Molecular Formula: C8H7F4NO Molecular Weight (g/mol): 209.144 MDL Number: MFCD04115895 InChI Key: MBNLPGBRZXRZOM-UHFFFAOYSA-N Synonym: 2-fluoro-5-trifluoromethoxy benzylamine,2-fluoro-5-trifluoromethoxy phenyl methanamine,2-fluoro-5-trifluoromethoxy phenyl methylamine,1-2-fluoro-5-trifluoromethoxy phenyl methanamine,2-fluoro-5-trifluoromethoxy benzenemethanamine,benzenemethanamine,2-fluoro-5-trifluoromethoxy PubChem CID: 3739302 IUPAC Name: [2-fluoro-5-(trifluoromethoxy)phenyl]methanamine SMILES: C1=CC(=C(C=C1OC(F)(F)F)CN)F