accessibility menu, dialog, popup

UserName

CAS RN 886362-07-8

H3CONHNH2N

CAS RN 886362-07-8

IUPAC Name: 4-methoxy-1H-indazol-3-amine
Synonyms: 4-methoxy-1H-indazol-3-amine
Molecular Weight (g/mol): 163.18
Molecular Formula: C8H9N3O
InChi Key: JZAMRGWQDJMEJD-UHFFFAOYSA-N
SMILES: COC1=C2C(N)=NNC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results