accessibility menu, dialog, popup

UserName

CAS RN 885269-54-5

CH3H3CSOONHOOHCH3CH3

CAS RN 885269-54-5

IUPAC Name: 1-(2,3,5,6-tetramethylbenzenesulfonamido)cyclohexane-1-carboxylic acid
Synonyms: 1-(2,3,5,6-tetramethylbenzenesulfonamido)cyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 339.45
Molecular Formula: C17H25NO4S
InChi Key: AUJXKRTUDQFEGI-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(=C1C)S(=O)(=O)NC1(CCCCC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results