accessibility menu, dialog, popup

UserName

CH3HNNF

CAS RN 884507-13-5

IUPAC Name: [(3-fluoropyridin-2-yl)methyl](methyl)amine
Synonyms: [(3-fluoropyridin-2-yl)methyl](methyl)amine
Molecular Weight (g/mol): 140.16
Molecular Formula: C7H9FN2
InChi Key: CRJRGUJNAQPYJG-UHFFFAOYSA-N
SMILES: CNCC1=C(F)C=CC=N1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results