missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-N-methylpyrid-2-ylmethylamine, Tech., Thermo Scientific™

Product Code. 10502484
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502484 1 g Amber Glass Bottle 1g
10265284 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10502484 Supplier Thermo Scientific Maybridge Supplier No. CC30946DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-13-5
Molecular Formula C7H9FN2
Molecular Weight (g/mol) 140.16
MDL Number MFCD09025827
InChI Key CRJRGUJNAQPYJG-UHFFFAOYSA-N
Synonym 3-fluoro-n-methylpyrid-2-ylmethylamine, 3-fluoropyridin-2-yl methyl methyl amine, 1-3-fluoropyridin-2-yl-n-methylmethanamine, 2-pyridinemethanamine, 3-fluoro-n-methyl, 2-pyridinemethanamine,3-fluoro-n-methyl, 3-fluoro 2-pyridyl methyl methylamine, 3-fluoro-n-methylpyrid-2-ylmethylamine,tech., 3-fluoro-n-methylpyridin-2-ylmethylamine
PubChem CID 18525718
IUPAC Name 1-(3-fluoropyridin-2-yl)-N-methylmethanamine
SMILES CNCC1=C(F)C=CC=N1

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 140.16
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Fluoro-N-methylpyrid-2-ylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.