accessibility menu, dialog, popup

UserName

CAS RN 870703-74-5

CH3H3CCH3OONNOOH3C

CAS RN 870703-74-5

IUPAC Name: tert-butyl 4-[2-(methoxycarbonyl)phenyl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[2-(methoxycarbonyl)phenyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 320.39
Molecular Formula: C17H24N2O4
InChi Key: JFZGCXORLWXLGH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1N1CCN(CC1)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results