accessibility menu, dialog, popup

UserName

CAS RN 86256-51-1

OFFFFBr

CAS RN 86256-51-1

IUPAC Name: 1-(bromomethyl)-3-fluoro-5-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-3-fluoro-5-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 273.03
Molecular Formula: C8H5BrF4O
InChi Key: PJLFSUVIEZRQRU-UHFFFAOYSA-N
SMILES: FC1=CC(CBr)=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results