missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-fluoro-5-(trifluoromethyl)pyridine, Thermo Scientific™

Product Code. 10452400
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10452400

Brand: Thermo Scientific Maybridge MAY00210DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 852062-17-0
Packaging Amber Glass Bottle
Molecular Formula C6H4F4N2
Quantity 1 g
Synonym 2-amino-3-fluoro-5-trifluoromethyl pyridine, 3-fluoro-5-trifluoromethyl pyridin-2-amine, 2-amino-3-fluoro-5-trifluoromethylpyridine, 3-fluoro-5-trifluoromethyl-2-pyridylamine, 3-fluoro-5-trifluoromethyl pyridine-2-amine, 3-fluoro-5-trifluoromethyl-2-pyridinamine
InChI Key HAVVQRHBCUBQGG-UHFFFAOYSA-N
SMILES C1=C(C=NC(=C1F)N)C(F)(F)F
IUPAC Name 3-fluoro-5-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol) 180.106
PubChem CID 25067349
Formula Weight 180.1
Percent Purity 97%
Chemical Name or Material 2-amino-3-fluoro-5-(trifluoromethyl)pyridine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.