CAS RN 84680-95-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1'-Bis(di-tert-butylphosphino)ferrocene, 98%, Thermo Scientific Chemicals
CAS: 84680-95-5 Molecular Formula: C26H44FeP2 Molecular Weight (g/mol): 474.43 MDL Number: MFCD01630818 InChI Key: DFRHTHSZMBROSH-UHFFFAOYSA-N IUPAC Name: 1,1'-Bis(di-tert-butylphosphino)ferrocene SMILES: [Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C