missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(di-tert-butylphosphino)ferrocene, 98%, Thermo Scientific Chemicals

Product Code. 11319705
Change view
Click to view available options
Quantity:
500 mg
2 g
Unit Size:
2g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319705 2 g 2g
11309705 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11319705 Supplier Thermo Scientific Alfa Aesar Supplier No. L19759.04
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

The rate of palladium-catalyzed amination of unactivated aryl chlorides is accelerated by sterically hindered chelating alkyl phosphines, ie, 1,1'-bis(di-tert-butylphosphino)ferrocene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The rate of palladium-catalyzed amination of unactivated aryl chlorides is accelerated by sterically hindered chelating alkyl phosphines, ie, 1,1′-bis(di-tert-butylphosphino)ferrocene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84680-95-5
Molecular Formula C26H44FeP2
Molecular Weight (g/mol) 474.43
MDL Number MFCD01630818
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
IUPAC Name 1,1'-Bis(di-tert-butylphosphino)ferrocene
SMILES [Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C

Specifications

Melting Point 73°C to 78°C
Quantity 2 g
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 474.43
Percent Purity 98%
Chemical Name or Material 1,1'-Bis(di-tert-butylphosphino)ferrocene
compliance-icons
Product Identifier
  • 1,1'-Bis(di-tert-butylphosphino)ferrocene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.