CAS RN 84504-69-8
IUPAC Name:
(2Z)-but-2-enedioic acid; 6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine
Synonyms:
irsogladine
Molecular Weight (g/mol):
372.16
Molecular Formula:
C13H11Cl2N5O4
InChi Key:
PJLVTVAIERNDEQ-BTJKTKAUSA-N
SMILES:
OC(=O)\C=C/C(O)=O.NC1=NC(=NC(N)=N1)C1=CC(Cl)=CC=C1Cl
1
–
1
of
1
results
1
Irsogladine Maleate, TRC
CAS: 84504-69-8 Molecular Formula: C9 H7 Cl2 N5 . C4 H4 O4 Molecular Weight (g/mol): 372.16 Synonym: 1,3,5-Triazine-2,4-diamine, 6-(2,5-dichlorophenyl)-, (2Z)-2-butenedioate (1:1) (9CI, ACI),1,3,5-Triazine-2,4-diamine, 6-(2,5-dichlorophenyl)-, (Z)-2-butenedioate (1:1) (ZCI),2,4-Diamino-6-(2,5-dichlorophenyl)-s-triazine maleate,Gaslon,Gaslon N,Irsogladine maleate,MN 1695,6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine (2Z)-2-butenedioate (1:1) IUPAC Name: (Z)-but-2-enedioic acid;6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine SMILES: Nc1nc(N)nc(n1)c2cc(Cl)ccc2Cl.OC(=O)\C=C/C(=O)O