missing translation for 'onlineSavingsMsg'
Learn More

Irsogladine Maleate, TRC

Product Code. 30405845
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405845 50 mg -
30394152 5 g 5g
30391688 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405845 Supplier Toronto Research Chemicals Supplier No. TRCI77700050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84504-69-8
Molecular Formula C9 H7 Cl2 N5 . C4 H4 O4
Molecular Weight (g/mol) 372.16
Synonym 1,3,5-Triazine-2,4-diamine, 6-(2,5-dichlorophenyl)-, (2Z)-2-butenedioate (1:1) (9CI, ACI), 1,3,5-Triazine-2,4-diamine, 6-(2,5-dichlorophenyl)-, (Z)-2-butenedioate (1:1) (ZCI), 2,4-Diamino-6-(2,5-dichlorophenyl)-s-triazine maleate, Gaslon, Gaslon N, Irsogladine maleate, MN 1695, 6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine (2Z)-2-butenedioate (1:1)
IUPAC Name (Z)-but-2-enedioic acid;6-(2,5-dichlorophenyl)-1,3,5-triazine-2,4-diamine
SMILES Nc1nc(N)nc(n1)c2cc(Cl)ccc2Cl.OC(=O)\C=C/C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H7Cl2N5.C4H4O4/c10-4-1-2-6(11)5(3-4)7-14-8(12)16-9(13)15-7;5-3(6)1-2-4(7)8/h1-3H,(H4,12,13,14,15,16);1-2H,(H,5,6)(H,7,8)/b;2-1-
Formula Weight 371.0188
Chemical Name or Material Irsogladine Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.