accessibility menu, dialog, popup

UserName

CAS RN 81067-38-1

ClClBrCl

CAS RN 81067-38-1

IUPAC Name: 1-bromo-2,3,5-trichlorobenzene
Synonyms: 1-bromo-2,3,5-trichlorobenzene
Molecular Weight (g/mol): 260.34
Molecular Formula: C6H2BrCl3
InChi Key: WOIGJFAVHOCDDW-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Cl)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results