accessibility menu, dialog, popup

UserName

CAS RN 81055-73-4

H3COOClCl

CAS RN 81055-73-4

IUPAC Name: ethyl 2,6-dichlorobenzoate
Synonyms: ethyl 2,6-dichlorobenzoate
Molecular Weight (g/mol): 219.06
Molecular Formula: C9H8Cl2O2
InChi Key: HAFAMHIZRCDTSS-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(Cl)C=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results