accessibility menu, dialog, popup

UserName

CAS RN 80653-69-6

H3COOSNN

CAS RN 80653-69-6

IUPAC Name: ethyl 2-[2-(pyridin-3-yl)-1,3-thiazol-4-yl]acetate
Synonyms: ethyl 2-[2-(pyridin-3-yl)-1,3-thiazol-4-yl]acetate
Molecular Weight (g/mol): 248.3
Molecular Formula: C12H12N2O2S
InChi Key: JGRCACXOIKCVIX-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CSC(=N1)C1=CN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results