accessibility menu, dialog, popup

UserName

CAS RN 7790-98-9

NH4OClOOO

CAS RN 7790-98-9

IUPAC Name: ammonium perchlorate
Synonyms: ammonium perchlorate
Molecular Weight (g/mol): 117.49
Molecular Formula: ClH4NO4
InChi Key: HHEFNVCDPLQQTP-UHFFFAOYSA-N
SMILES: [NH4+].[O-][Cl](=O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results