missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Perchlorate Solution, Solstice

Product Code. 15658300 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15658300 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15658300 Supplier Honeywell Research Chemicals Supplier No. 099112.5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-98-9
Molecular Formula ClH4NO4
Molecular Weight (g/mol) 117.485
MDL Number MFCD00011421
InChI Key HHEFNVCDPLQQTP-UHFFFAOYSA-N
PubChem CID 24639
IUPAC Name azanium;perchlorate
SMILES [NH4+].[O-]Cl(=O)(=O)=O

Specifications

Quantity 2.5 L
Linear Formula NH4ClO4
UN Number 3211
Formula Weight 117.49g/mol
Grade Reagent
Chemical Name or Material Ammonium Perchlorate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.