CAS RN 77573-43-4
CAS RN 77573-43-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ingenol-5,20-acetonide, MedChemExpress
MedChemExpress Ingenol-5,20-acetonide is an intermediate from ingenol for synthesis of ingenoids; improved stability compared to ingenol.