accessibility menu, dialog, popup

UserName

CAS RN 774549-97-2

NNNClSOOCl

CAS RN 774549-97-2

IUPAC Name: 7-chloro-4-[4-(4-chlorobenzenesulfonyl)piperazin-1-yl]quinoline
Synonyms: 7-chloro-4-[4-(4-chlorobenzenesulfonyl)piperazin-1-yl]quinoline
Molecular Weight (g/mol): 422.32
Molecular Formula: C19H17Cl2N3O2S
InChi Key: GIEHIZKCIZLXLF-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)(=O)N1CCN(CC1)C1=CC=NC2=CC(Cl)=CC=C12
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results