missing translation for 'onlineSavingsMsg'
Learn More

KM11060, MedChemExpress

Product Code. 16497766
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497766 100 mg 100mg
16487988 10 mg 10mg
16453917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497766 Supplier Medchem Express Supplier No. HY19970
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress KM11060 is a corrector of the F508 deletion (F508del)-cystic fibrosis transmembrane conductance regulator (CFTR) trafficking defect. KM11060 can be used for the research of F508del-CFTR processing defect and development of cystic fibrosis therapeutics.

KM11060 is a corrector of the F508 deletion (F508del)-cystic fibrosis transmembrane conductance regulator (CFTR) trafficking defect. KM11060 can be used for the research of F508del-CFTR processing defect and development of cystic fibrosis therapeutics.

Specifications

Chemical Name or Material KM11060
CAS 774549-97-2
Color White
Purity Grade Notes Research
Molecular Formula C19H17Cl2N3O2S
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (118.39 mM; Need ultrasonic)
SMILES O=S(N1CCN(C2=CC=NC3=CC(Cl)=CC=C23)CC1)(C4=CC=C(Cl)C=C4)=O
Molecular Weight (g/mol) 422.33
Formula Weight 422.33
Percent Purity 99.59%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.